Learn More
TARGETMOL CHEMICALS INC KHK-IN-2 25MG
SDP
Catalog No. 502581513
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KHK-IN-2 is a selective inhibitor of ketohexokinase with an IC50 value of 0.45 uM. purity: 98%
Catalog No. 50-258-1513 Supplier TARGETMOL CHEMICALS INC Supplier No. T1175725MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KHK-IN-2 is a selective inhibitor of ketohexokinase with an IC50 value of 0.45 uM. purity: 98%