Learn More
Medchemexpress LLC HY-103478 5mg Medchemexpress, KHK-IN-2 CAS:2135304-43-5 Purity:>98%
SDP
Catalog No. 501968120
Encompass_Preferred
Medchemexpress, HY-103478 5mg KHK-IN-2 CAS:2135304-43-5 KHK-IN-2 is a potent and selective ketohexokinase (KHK) inhibitor with an IC50 of 0.45 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8120 Supplier Medchemexpress LLC Supplier No. HY1034785MG
Medchemexpress, HY-103478 5mg KHK-IN-2 CAS:2135304-43-5 KHK-IN-2 is a potent and selective ketohexokinase (KHK) inhibitor with an IC50 of 0.45 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.