Learn More
TARGETMOL CHEMICALS INC KHS 101 10MG
SDP
Catalog No. 502878915
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. KHS101 is a novel inhibitor of transforming acidic coiled-coil protein 3 (TACC3). It is a selective inducer of neuronal differentiation. Purity 98%
Catalog No. 50-287-8915 Supplier TARGETMOL CHEMICALS INC Supplier No. T496810MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. KHS101 is a novel inhibitor of transforming acidic coiled-coil protein 3 (TACC3). It is a selective inducer of neuronal differentiation. Purity 98%