Learn More
Medchemexpress LLC KHS101 hydrochloride | 1784282-12-7 | 99.65% | 375.92 | 100 MG
SDP
Catalog No. 5000344989
Encompass_Preferred
KHS101 hydrochloride is a selective neuronal differentiation inducer that can cross the blood-brain barrier. It exhibits selective cytotoxicity against glioblastoma multiforme (GBM) and interacts with transforming acidic coiled-coil protein (TACC3). This compound is suitable for GBM research.
  • Selective neuronal differentiation inducer
  • Crosses the blood-brain barrier
  • Selective cytotoxicity against glioblastoma multiforme (GBM)
  • Interacts with transforming acidic coiled-coil protein (TACC3)
  • Useful in GBM research
Catalog No. 50-003-44989 Supplier Medchemexpress LLC Supplier No. HY10996A10MM/1ML
KHS101 hydrochloride is a selective neuronal differentiation inducer that can cross the blood-brain barrier. It exhibits selective cytotoxicity against glioblastoma multiforme (GBM) and interacts with transforming acidic coiled-coil protein (TACC3). This compound is suitable for GBM research.
  • Selective neuronal differentiation inducer
  • Crosses the blood-brain barrier
  • Selective cytotoxicity against glioblastoma multiforme (GBM)
  • Interacts with transforming acidic coiled-coil protein (TACC3)
  • Useful in GBM research