Learn More
Diagnostic Biosystems KI-67 Ki-67
Catalog No. 502407425
Encompass_Preferred
This antibody reacts with a nuclear antigen present in proliferating human cells. It can be used to evaluate the Ki-67 labeling index in various tumors.
Catalog No. 50-240-7425 Supplier Diagnostic Biosystems Supplier No. PDR048
This antibody reacts with a nuclear antigen present in proliferating human cells. It can be used to evaluate the Ki-67 labeling index in various tumors.