Learn More
Diagnostic Biosystems KI-67 ANTIGEN Ki-67 Antigen
Catalog No. 502407560
Encompass_Preferred
Ki-67 is a nuclear protein, which is expressed in proliferating cells. Ki-67 is preferentially expressed during late G1-, S-, M-, and G2-phases of the cell cycle, while cells in the G0 (quiescent) phase are negative for this protein.
Catalog No. 50-240-7560 Supplier Diagnostic Biosystems Supplier No. RMPD004
Ki-67 is a nuclear protein, which is expressed in proliferating cells. Ki-67 is preferentially expressed during late G1-, S-, M-, and G2-phases of the cell cycle, while cells in the G0 (quiescent) phase are negative for this protein.