Learn More
Biogenex Laboratories Ki-67 : Ki88 : 6mL
SDP
Supplier:  Biogenex Laboratories AM3705M
Encompass_Preferred
Monoclonal antibody Ki88 reacts with a human nuclear antigen expressedin proliferating cells but absent in resting cells. It is expressed during allphases of the cell cycle except Go. Ki-67 antigen is a potent tool for rapidevaluation of the growth fraction of any given human cell subset.Immunostaining of Ki-67 antigen in normal tissue shows nuclearreactivity in cells of germinal centers of cortical follicles, corticalthymocytes, neck cells of gastrointestinal mucosa, and undifferentiatedspermatogonia. Resting cells such as lymphocytes, monocytes, parietalcells and Paneth’s cells of gastrointestinal mucosa, hepatocytes, renalcells, and mature sperm cells do not stain
Catalog No. 50-197-8847