Learn More
AVANTOR MASTERFLEX (VWR LLC) Key Instruments FR4A41BVBN-D Valved Acrylic Flowmeter for gases, 1 to 10 LPM, 100 mm Scale
Catalog No. NC1487166
Encompass
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters' inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is a tripod base (32462-50 sold separately) which allows for mobility from bench to bench. Note: There are many additional types of acrylic flowmeters not listed here. Contact our Application Specialits for quotes on acrylic meters with special requirements.
Catalog No. NC1487166 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3247052
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters' inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is a tripod base (32462-50 sold separately) which allows for mobility from bench to bench. Note: There are many additional types of acrylic flowmeters not listed here. Contact our Application Specialits for quotes on acrylic meters with special requirements.