Learn More
Apexbio Technology LLC Ki16198, 10mg. Cas: 355025-13-7 MFCD: MFCD23704182. LPA antagonist
SDP
Supplier:  Apexbio Technology LLC B159110
Encompass_Preferred
Ki16198 is the methyl ester of Ki16425. Ki16198, a LPA antagonist, inhibits LPA1- and LPA3-induced inositol phosphate production with the Ki of 0.34 M and 0.93 M, respectively, shows weaker inhibition for LPA2, no activity at LPA4, LPA5, LPA6[1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5269