Learn More
Apexbio Technology LLC Ki16198 355025-13-7 200mg
SDP
Catalog No. 5000062485
Encompass_Preferred
Ki16198 (CAS 355025-13-7) is the methyl ester derivative of Ki16425 and functions as an antagonist of lysophosphatidic acid (LPA) receptors preferentially inhibiting LPA1 and LPA3 receptor subtypes Ki16198 reduces receptor-mediated inositol phosphate production with Ki values of 0 34 M (LPA1) and 0 93 M (LPA3) showing minimal inhibition of LPA2 and no significant activity against LPA4 6 In vitro Ki16198 attenuates LPA-induced proliferation migration invasion and proMMP-9 expression in cancer cells In vivo studies report suppression of pancreatic tumor growth metastasis and ascites formation highlighting its utility in investigating LPA-related cancer progression pathways
Catalog No. 50-000-62485 Supplier Apexbio Technology LLC Supplier No. B1591200
Ki16198 (CAS 355025-13-7) is the methyl ester derivative of Ki16425 and functions as an antagonist of lysophosphatidic acid (LPA) receptors preferentially inhibiting LPA1 and LPA3 receptor subtypes Ki16198 reduces receptor-mediated inositol phosphate production with Ki values of 0 34 M (LPA1) and 0 93 M (LPA3) showing minimal inhibition of LPA2 and no significant activity against LPA4 6 In vitro Ki16198 attenuates LPA-induced proliferation migration invasion and proMMP-9 expression in cancer cells In vivo studies report suppression of pancreatic tumor growth metastasis and ascites formation highlighting its utility in investigating LPA-related cancer progression pathways