Learn More
Selleck Chemical LLC Ki16425 S1315-2mg
SDP
Catalog No. 503108344
Encompass_Preferred
Ki16425 is a competitive potent and reversible antagonist to LPA1 LPA2 and LPA3 with Ki of 0 34 M 6 5 M and 0 93 M in RH7777 cell lines respectively shows no activity at LPA4 LPA5 LPA6
Catalog No. 50-310-8344 Supplier Selleck Chemical LLC Supplier No. S13152MG
Ki16425 is a competitive potent and reversible antagonist to LPA1 LPA2 and LPA3 with Ki of 0 34 M 6 5 M and 0 93 M in RH7777 cell lines respectively shows no activity at LPA4 LPA5 LPA6