Learn More
Medchemexpress LLC HY-13285 10mg , Ki16425 CAS:355025-24-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1328510MG
Encompass_Preferred
HY-13285 10mg Ki16425 CAS: 355025-24-0 Ki16425 (Debio 0719) is a subtype-selective, competitive antagonist of the EDG-family receptors, LPA1 and LPA3 with Kis of 0.34 μM and 0.93 μM, respectively. Ki16425 (Debio 0719) reduces the LPA-induced activation of p42/p44 MAPK. Ki16425 can also inhibit LPA-induced dephosphorylation of Yes-associated protein (YAP)/TAZ in HEK293A cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9693