Learn More
Selleck Chemical LLC Ki16425 5mg 355025-24-0
SDP
Catalog No. 508180
Encompass_Preferred
Ki16425 is a competitive, potent and reversible antagonist to LPA1, LPA2 and LPA3 with Ki of 0.34 ?M, 6.5 ?M and 0.93 ?M in RH7777 cell lines, respectively, shows no activity at LPA4, LPA5, LPA6. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-818-0 Supplier Selleck Chemical LLC Supplier No. S13155MG
Add to Cart
Add to Cart
Ki16425 is a competitive, potent and reversible antagonist to LPA1, LPA2 and LPA3 with Ki of 0.34 ?M, 6.5 ?M and 0.93 ?M in RH7777 cell lines, respectively, shows no activity at LPA4, LPA5, LPA6. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.