Learn More
Selleck Chemical LLC Ki20227
SDP
Catalog No. 5000003020
Encompass_Preferred
Ki20227 is an orally active and highly selective inhibitor of c-Fms tyrosine kinase(CSF1R) with IC50 of 2 nM 12 nM 451 nM and 217 nM for c-Fms vascular endothelial growth factor receptor-2 (KDR/VEGFR-2) stem cell factor receptor (c-Kit) and platelet-derived growth factor receptor beta (PDGFR ) respectively
Catalog No. 50-000-03020 Supplier Selleck Chemical LLC Supplier No. S76882MG
Ki20227 is an orally active and highly selective inhibitor of c-Fms tyrosine kinase(CSF1R) with IC50 of 2 nM 12 nM 451 nM and 217 nM for c-Fms vascular endothelial growth factor receptor-2 (KDR/VEGFR-2) stem cell factor receptor (c-Kit) and platelet-derived growth factor receptor beta (PDGFR ) respectively