Learn More
Medchemexpress LLC HY-101140 1mg Medchemexpress, KI696 CAS:1799974-70-1 Purity:>98%
SDP
Catalog No. 502028648
Encompass_Preferred
Medchemexpress, HY-101140 1mg KI696 CAS:1799974-70-1 KI696 is a high affinity probe that disrupts the Keap1/NRF2 interaction. KI696 is a potent and selective inhibitor of the KEAP1/NRF2 interaction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8648 Supplier Medchemexpress LLC Supplier No. HY1011401MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-101140 1mg KI696 CAS:1799974-70-1 KI696 is a high affinity probe that disrupts the Keap1/NRF2 interaction. KI696 is a potent and selective inhibitor of the KEAP1/NRF2 interaction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.