Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KI696 isomer | 1799974-69-8 | 99.3% | 550.63 | C28H30N4O6S | 25 MG
SDP

Catalog No. 5000213534
Encompass_Preferred

KI696 isomer is the less active stereoisomer of KI696, a high-affinity probe that disrupts the Keap1-NRF2 interaction. Supplied as a white to off-white solid for research use, it is offered in solid and DMSO solution formats; follow recommended storage conditions for stability.

  • Less active stereoisomer that disrupts the Keap1-NRF2 interaction.
  • High purity (99.3%).
  • White to off-white solid physical form.
  • Molecular formula C28H30N4O6S; molecular weight 550.63.
  • Available as solids and 10 mM solutions in DMSO.
  • Store powder at -20°C and solutions at -80°C for long-term stability.

Catalog No. 50-002-13534 Supplier Medchemexpress LLC Supplier No. HY101140A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KI696 isomer is the less active stereoisomer of KI696, a high-affinity probe that disrupts the Keap1-NRF2 interaction. Supplied as a white to off-white solid for research use, it is offered in solid and DMSO solution formats; follow recommended storage conditions for stability.

  • Less active stereoisomer that disrupts the Keap1-NRF2 interaction.
  • High purity (99.3%).
  • White to off-white solid physical form.
  • Molecular formula C28H30N4O6S; molecular weight 550.63.
  • Available as solids and 10 mM solutions in DMSO.
  • Store powder at -20°C and solutions at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.