Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KI696 isomer | 1799974-69-8 | 99.3% | 550.63 g/mol | C28H30N4O6S | 1 ML
SDP

Supplier:  Medchemexpress LLC HY101140A10MM 1ML

Encompass_Preferred

KI696 isomer is the less active stereoisomer of a high-affinity small-molecule probe that disrupts the Keap1-NRF2 protein-protein interaction. It is provided as a ready-to-use solution for biochemical and cell-based research exploring modulation of the KEAP1-NRF2 pathway.

  • Less active stereoisomer useful as a comparator or control in Keap1-NRF2 studies.
  • Supplied as a 10 mM solution in DMSO (1 mL vial) ready for immediate use.
  • High purity (≈99.3%) suitable for biochemical and cell-based assays.
  • Well characterized with molecular weight 550.63 g/mol and CAS 1799974-69-8.
  • Stable under recommended storage conditions to preserve compound integrity.

Catalog No. 50-002-13536


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.