Learn More
Medchemexpress LLC HY-12038 10mg Medchemexpress, Ki8751 CAS:228559-41-9 Purity:>98%
SDP
Catalog No. 502080938
Encompass_Preferred
Medchemexpress, HY-12038 10mg Ki8751 CAS:228559-41-9 Ki8751 is a potent VEGFR2 inhibitor with an IC50 of 0.9 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0938 Supplier Medchemexpress LLC Supplier No. HY1203810MG
Medchemexpress, HY-12038 10mg Ki8751 CAS:228559-41-9 Ki8751 is a potent VEGFR2 inhibitor with an IC50 of 0.9 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.