Learn More
Aviva Systems Biology Corp PUM3 Antibody - N-terminal region (ARP40336_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40336_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA0020. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human KIAA0020. Alternate names: PEN, PUF6, XTP5, HLA-HA8, RP11-526D20.2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Pig, Rabbit, Rat
Catalog No. 50-014-53191