Learn More
Aviva Systems Biology Corp KIAA0040 Antibody - middle region (ARP39121_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39121_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA0040. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KIAA0040. Alternate names: MGC133301ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56913