Learn More
Aviva Systems Biology Corp PCLAF Antibody - N-terminal region (ARP46033_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46033_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA0101. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human KIAA0101. Alternate names: L5, NS5ATP9, OEATC-1, OEATC1, PAF, p15(PAF), OEATC, p15PAF, p15/PAFApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-237