Learn More
Aviva Systems Biology Corp KIAA0152 Antibody - C-terminal region (ARP46201_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46201_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA0152. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human KIAA0152. Alternate names: KIAA0152ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-244