Learn More
Aviva Systems Biology Corp RUBCN Antibody - middle region (ARP58950_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58950_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA0226. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human KIAA0226. Alternate names: RUBICONApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-537