Learn More
Aviva Systems Biology Corp SUSD6 Antibody - N-terminal region (ARP47062_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47062_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA0247. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KIAA0247. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-59462
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.