Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ KIAA0261 Recombinant Protein

Catalog No. p-3719771 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-012-294 10 μg
89-012-295 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-012-294 Supplier Abnova™ Supplier No. H00023063P0110
Only null left
Add to Cart
Add to Cart

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH17393
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 23063
Molecular Weight (g/mol) 69.96
Name KIAA0261 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MDLDRASLDLMIRLLELEQDASSAKLLNEKDMNKIKEKIRRLCETVHNKHLDLENITTGHLAMETLLSLTSKRAGDWFKEELRLLGGLDHIVDKVKECVDHLSRDEDEEKLVASLWGAERCLRVLESVTVHNPENQSYLIAYKDSQLIVSSAKALQHCEELIQQYNRAEDSICLADSKPLPHQNVTNHVGKAVEDCMRAIIGVLLNLTNDNEWGSTKTGEQDGLIGTALNCVLQVPKYLPQEQRFDIRVLLFLERERAAQLAESKTDELIKDAPTTQHDKSGEWQETSGEIQWVSTEKTDGTEEKHKKEEEDEELDLNKALQHAGKHMEDCIVASYTALLLGCLCQESPINVTTVREYLPEGDFSIMTEMLKKFLSFMNLTCAVGTTGQKSISRVIEYLEHC
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias FOE/KIAA0261/WAPL
Common Name WAPAL
Gene Symbol WAPAL
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.