Learn More
Aviva Systems Biology Corp KIAA0317 Antibody - middle region (ARP44875_P050) | 100uL
SDP
Catalog No. 5001458924
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA0317. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KIAA0317. Alternate names: KIAA0317, ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-58924 Supplier Aviva Systems Biology Corp Supplier No. ARP44875_P050
This is a rabbit polyclonal antibody against KIAA0317. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KIAA0317. Alternate names: KIAA0317, ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish