Learn More
Aviva Systems Biology Corp KIAA0317 Antibody - N-terminal region (ARP44874_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44874_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA0317. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KIAA0317. Alternate names: KIAA0317, ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58923