Learn More
Aviva Systems Biology Corp KIAA0319 Antibody - N-terminal region (ARP47083_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47083_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA0319. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human KIAA0319. Alternate names: DLX2, DYLX2, DYX2, MGC176717ApplicationIHC, WB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-53503