Learn More
Aviva Systems Biology Corp KIAA0737 Antibody - N-terminal region (ARP39144_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39144_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA0737. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KIAA0737. Alternate names: LCP1, MIG7, C14orf92, KIAA0737ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56920