Learn More
Aviva Systems Biology Corp KIAA0776 Antibody - middle region (ARP55228_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55228_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA0776. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KIAA0776. Alternate names: RP3-393D12.1, NLBP, RCAD, Maxer, KIAA0776ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61440