Learn More
Aviva Systems Biology Corp EEF1AKNMT Antibody - C-terminal region (ARP41852_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41852_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA0859. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human KIAA0859. Alternate names: 5630401D24Rik, CGI-01, FLJ10310, feat, KIAA0859ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58018