Learn More
Aviva Systems Biology Corp KHDC4 Antibody - middle region (ARP55116_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55116_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA0907. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KIAA0907. Alternate names: RP11-336K24.1, BLOM7ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53916