Learn More
Aviva Systems Biology Corp KIAA0999 Antibody - middle region (ARP53784_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53784_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA0999. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human KIAA0999. Alternate names: FLJ12240, KIAA0999, L19, QSK, SIK-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-887