Learn More
Aviva Systems Biology Corp KIAA1191 Antibody - middle region (ARP56290_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56290_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA1191. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KIAA1191. Alternate names: FLJ21022, p60MONOXApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-014-61798