Learn More
Aviva Systems Biology Corp CEMIP Antibody - middle region (ARP42526_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42526_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA1199. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KIAA1199. Alternate names: TMEM2L, CCSP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-58255