Learn More
Aviva Systems Biology Corp KIAA1267 Antibody - N-terminal region (ARP55255_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55255_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA1267. It was validated on Western Blot. . Alternate names: DKFZp686P06109, DKFZp727C091, MGC102843, NSL1, MSL1v1, CENP-36, KIAA1267ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-018