Learn More
Aviva Systems Biology Corp KIF1BP Antibody - C-terminal region (ARP62341_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62341_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA1279. It was validated on Western Blot. . Alternate names: DKFZp586B0923, KBP, TTC20ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-654