Learn More
Aviva Systems Biology Corp KIAA1324 Antibody - N-terminal region (ARP49563_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49563_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA1324. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KIAA1324. Alternate names: EIG121, MGC150624, RP11-352P4.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53590