Learn More
Sino Biological Recombinant Human BRAF/B-Raf Protein (d486-490, V471F, GST Tag)
Catalog No. 502575803
Encompass_Preferred
Recombinant human BRAF (d486-490 V471F) (381-end or exon 9-18) protein was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5803 Supplier Sino Biological Supplier No. B0812LG50
Recombinant human BRAF (d486-490 V471F) (381-end or exon 9-18) protein was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.