Learn More
Aviva Systems Biology Corp KIAA1604 Antibody - C-terminal region (ARP40957_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40957_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA1604. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human KIAA1604. Alternate names: EIF4GL, KIAA1604, NCM, fSAPbApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-284-904