Learn More
Aviva Systems Biology Corp MEAK7 Antibody - middle region (ARP57504_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57504_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA1609. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human KIAA1609. Alternate names: MGC25024, KIAA1609ApplicationWB. Species reactivity: Human
Catalog No. 50-286-324