Learn More
Aviva Systems Biology Corp KIAA1618 Antibody - N-terminal region (ARP36307_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36307_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA1618. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KIAA1618. Alternate names: ALO17, MYMY2, MYSTR, NET57, C17orf27, KIAA1618ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56111