Learn More
Aviva Systems Biology Corp KIAA1754L Antibody - C-terminal region (ARP44575_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44575_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA1754L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human KIAA1754L. Alternate names: KIAA1754LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-58847