Learn More
Sigma Aldrich Fine Chemicals Biosciences KiCqStartR Probe qPCR Read
Catalog No. 502914257
Encompass
KiCqStart Probe qPCR ReadyMix isanadvanced qPCR reagent system for both fast and conventional PCR cycling protocols or instruments. It is a versatile and robust SOLUTIONon that provides the ultimate sensitivity and high PCR efficiency using a variety of fluorogenic probe chemistries including TaqMan(R) hydrolysis probes. KiCqStart Probe qPCR ReadyMix is provided as a 2X concentrated ready-to-use reaction cocktail that contains all required reaction components except primers probe(s) and DNA template. The light blue color of an inert tracer dye simplifies reaction assembly in white or clear plates and helps to minimize pipetting or mixing errors. It does not interfere with qPCR performance or affect the stability of the product.A key component of KiCqStart Probe qPCR ReadyMix is an ultra-pure processive thermostable DNA polymerase that is free of detectable E. coli DNAA§. KiCqStart Probe qPCR ReadyMix is ideal for demanding qPCR applications such as bacterial pathogen detection w
Catalog No. 50-291-4257 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. KCQS061250RXN
KiCqStart Probe qPCR ReadyMix isanadvanced qPCR reagent system for both fast and conventional PCR cycling protocols or instruments. It is a versatile and robust SOLUTIONon that provides the ultimate sensitivity and high PCR efficiency using a variety of fluorogenic probe chemistries including TaqMan(R) hydrolysis probes. KiCqStart Probe qPCR ReadyMix is provided as a 2X concentrated ready-to-use reaction cocktail that contains all required reaction components except primers probe(s) and DNA template. The light blue color of an inert tracer dye simplifies reaction assembly in white or clear plates and helps to minimize pipetting or mixing errors. It does not interfere with qPCR performance or affect the stability of the product.A key component of KiCqStart Probe qPCR ReadyMix is an ultra-pure processive thermostable DNA polymerase that is free of detectable E. coli DNAA§. KiCqStart Probe qPCR ReadyMix is ideal for demanding qPCR applications such as bacterial pathogen detection w