Learn More
Enterprise Technology Solutions Kidde Fire And Safety Fire Extinguisher, 5B:C, 3-1/4"Wx3-1/4"Lx11-7/10"H, White
SDP
Catalog No. 5000828166
Encompass_Preferred
Designed for single use, this compact extinguisher contains a fast-acting extinguishing agent to quickly put out flames during an emergency. Easy-to-read gauge lets you quickly see if the extinguisher is charged and ready for use. Clear instruction label features descriptive graphics to show you the steps required to operate. Easy-to-pull safety pin allows quick use during emergencies. Metal handle is rust-resistant and impact-resistant for reliable performance. Extinguisher is UL rated 5B:C and is suitable for use on liquid and energized electrical fires.
Catalog No. 50-008-28166 Supplier Enterprise Technology Solutions Supplier No. KID21006287MTL
Designed for single use, this compact extinguisher contains a fast-acting extinguishing agent to quickly put out flames during an emergency. Easy-to-read gauge lets you quickly see if the extinguisher is charged and ready for use. Clear instruction label features descriptive graphics to show you the steps required to operate. Easy-to-pull safety pin allows quick use during emergencies. Metal handle is rust-resistant and impact-resistant for reliable performance. Extinguisher is UL rated 5B:C and is suitable for use on liquid and energized electrical fires.