Learn More
MSC Kidde Mounting Bracket, 4-5 lb Extinguisher, Stainless Steel, 1 ea
Catalog No. 5000782162
Encompass
This bracket is designed to securely mount a 4-5 lb fire extinguisher for use in homes, boats, or vehicles, ensuring easy access when needed.
  • Designed to mount 4-5 lb fire extinguishers for quick access
  • Easy to install with four 1/4 inch screws or bolts (not included)
  • DOT and USCG approved when used with corresponding UL/USCG approved fire extinguisher models
  • Features a non-corrosive stainless steel strap for rugged applications
  • Compatible with dry chemical extinguisher types
  • Designed for wall mounting applications
Catalog No. 50-007-82162 Supplier MSC Supplier No. 04034286
This bracket is designed to securely mount a 4-5 lb fire extinguisher for use in homes, boats, or vehicles, ensuring easy access when needed.
  • Designed to mount 4-5 lb fire extinguishers for quick access
  • Easy to install with four 1/4 inch screws or bolts (not included)
  • DOT and USCG approved when used with corresponding UL/USCG approved fire extinguisher models
  • Features a non-corrosive stainless steel strap for rugged applications
  • Compatible with dry chemical extinguisher types
  • Designed for wall mounting applications