Learn More
MSC Kidde Vehicle Metal Bracket, 10 Lb Dry Chemical, 1 ea
Catalog No. 5000794359
Encompass
This vehicle-mount fire extinguisher bracket is designed for securely mounting 10 lb dry chemical fire extinguishers. It is suitable for wall-mounting applications and adheres to U.S.C.G. standards for reliable installation.
  • Vehicle-mount fire extinguisher bracket
  • Wall mounting application
  • Maximum extinguisher capacity: 10 lb
  • Compatible with dry chemical extinguishers
  • Meets U.S.C.G. standards
  • Country of origin: China
Catalog No. 50-007-94359 Supplier MSC Supplier No. 57000077
This vehicle-mount fire extinguisher bracket is designed for securely mounting 10 lb dry chemical fire extinguishers. It is suitable for wall-mounting applications and adheres to U.S.C.G. standards for reliable installation.
  • Vehicle-mount fire extinguisher bracket
  • Wall mounting application
  • Maximum extinguisher capacity: 10 lb
  • Compatible with dry chemical extinguishers
  • Meets U.S.C.G. standards
  • Country of origin: China