Learn More
Discovery Life Sciences Kidney Cancer Active Tx PBMC
SDP
Catalog No. 5000803975
Encompass
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays
Catalog No. 50-008-03975 Supplier Discovery Life Sciences Supplier No. 120580
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays