Learn More
StatLab Kidney IHC High Quality Control Slides
Description
- Use these slides with confidence to evaluate stain quality and performance in histology laboratories
- StatLab control slides are created using Millennia adhesion white glass slides with 90° corners
- Each package includes specific staining protocol
- 25/package, stained slide is not included
- Tissue: Kidney
Specifications
Specifications
| Product Type | Control Slide |