Learn More
Meso Scale Discovery Kidney Injury PNL 1 rat Kit
SDP
Catalog No. 5000080559
Encompass
The Meso Scale Discovery Kidney Injury Panel 1 Rat Kit is designed to measure biomarkers of kidney injury in rat serum plasma and urine samples The kit contains a 96-well plate coated with capture antibodies specific for kidney injury biomarkers KIM-1 NGAL Clusterin Cystatin-C and Osteopontin The assay uses electrochemiluminescence detection technology for high sensitivity and broad dynamic range The kit includes all necessary reagents and calibrators for quantitative analysis of the 5 biomarkers The kit is for research use only and is not intended for diagnostic procedures The kit allows for high-throughput analysis of kidney injury biomarkers in preclinical rat studies
Catalog No. 50-000-80559 Supplier Meso Scale Discovery Supplier No. K15162C4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Kidney Injury Panel 1 Rat Kit is designed to measure biomarkers of kidney injury in rat serum plasma and urine samples The kit contains a 96-well plate coated with capture antibodies specific for kidney injury biomarkers KIM-1 NGAL Clusterin Cystatin-C and Osteopontin The assay uses electrochemiluminescence detection technology for high sensitivity and broad dynamic range The kit includes all necessary reagents and calibrators for quantitative analysis of the 5 biomarkers The kit is for research use only and is not intended for diagnostic procedures The kit allows for high-throughput analysis of kidney injury biomarkers in preclinical rat studies